siRNA Details
| Gene Symbol: RGD1359634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007708.1 | NP_001007709.1 | 4 | 630 | ||
| XM_006241992.2 | XP_006242054.1 | 5 | 477 | |||
| XM_008765713.1 | XP_008763935.1 | 6 | 503 | |||
| GenBank mRNA | BC079039.1 | - | 4 | 630 | 7114722 | |

RGD1359634
Genomic Map