siRNA Details
| Gene Symbol: Kdelr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127546.1 | NP_001121018.1 | 3 | 406 | ||
| XM_008765714.1 | XP_008763936.1 | 3 | 1124 | |||
| GenBank mRNA | BC162033.1 | - | 3 | 406 | 9103157 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelr3| Gene Symbol: Kdelr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127546.1 | NP_001121018.1 | 3 | 406 | ||
| XM_008765714.1 | XP_008763936.1 | 3 | 1124 | |||
| GenBank mRNA | BC162033.1 | - | 3 | 406 | 9103157 | |