siRNA Details
| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015018.1 | NP_001015018.1 | 1 | 301 | ||
| GenBank mRNA | BC092629.1 | - | 1 | 301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx17| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015018.1 | NP_001015018.1 | 1 | 301 | ||
| GenBank mRNA | BC092629.1 | - | 1 | 301 | ||