siRNA Details
| Gene Symbol: Mkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750385.3 | XP_003750433.1 | 12 | 8069 | ||
| XM_006242130.2 | XP_006242192.1 | 13 | 3126 | |||
| XM_008765806.1 | XP_008764028.1 | 12 | 3739 | |||
| XM_235497.8 | XP_235497.6 | 13 | 2800 | |||
| GenBank mRNA | AB588919.1 | - | 13 | 2687 | ||
| AB588920.1 | - | 12 | 2542 | |||
| AB588921.1 | - | 13 | 2669 | |||

Mkl1
Genomic Map