siRNA Details
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130504.1 | NP_001123976.1 | 3 | 275 | ||
XM_006242086.2 | XP_006242148.1 | 3 | 540 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130504.1 | NP_001123976.1 | 3 | 275 | ||
XM_006242086.2 | XP_006242148.1 | 3 | 540 |