siRNA Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130504.1 | NP_001123976.1 | 3, 4 | 297 | ||
| XM_006242086.2 | XP_006242148.1 | 3, 4 | 562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpm| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130504.1 | NP_001123976.1 | 3, 4 | 297 | ||
| XM_006242086.2 | XP_006242148.1 | 3, 4 | 562 | |||