siRNA Details
| Gene Symbol: Ndufa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130505.1 | NP_001123977.1 | 2 | 188 | ||
| GenBank mRNA | FQ218741.1 | - | 2 | 181 | ||
| FQ223548.1 | - | 2 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufa6| Gene Symbol: Ndufa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130505.1 | NP_001123977.1 | 2 | 188 | ||
| GenBank mRNA | FQ218741.1 | - | 2 | 181 | ||
| FQ223548.1 | - | 2 | 176 | |||