siRNA Details
| Gene Symbol: Mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191788.1 | NP_001178717.1 | 3 | 670 | ||
| XM_006242091.1 | XP_006242153.1 | 4 | 931 | |||
| GenBank mRNA | FQ232982.1 | - | 3 | 669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcat| Gene Symbol: Mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191788.1 | NP_001178717.1 | 3 | 670 | ||
| XM_006242091.1 | XP_006242153.1 | 4 | 931 | |||
| GenBank mRNA | FQ232982.1 | - | 3 | 669 | ||