siRNA Details
| Gene Symbol: Scube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134884.1 | NP_001128356.1 | 21 | 2837 | ||
| XM_006242092.2 | XP_006242154.1 | 20 | 2797 | |||
| XM_008765721.1 | XP_008763943.1 | 19 | 2907 | |||
| GenBank mRNA | BC086354.1 | - | 2 | 287 | ||

Scube1
Genomic Map