siRNA Details
| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108103.1 | NP_001101573.1 | 10 | 2734 | ||
| XM_006242194.2 | XP_006242256.1 | 10 | 3123 | |||
| XM_006242196.2 | XP_006242258.1 | 10 | 2730 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brd1| Gene Symbol: Brd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108103.1 | NP_001101573.1 | 10 | 2734 | ||
| XM_006242194.2 | XP_006242256.1 | 10 | 3123 | |||
| XM_006242196.2 | XP_006242258.1 | 10 | 2730 | |||