siRNA Details
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108105.1 | NP_001101575.1 | 5 | 914 | ||
XM_006242201.2 | XP_006242263.1 | 6 | 776 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108105.1 | NP_001101575.1 | 5 | 914 | ||
XM_006242201.2 | XP_006242263.1 | 6 | 776 |