siRNA Details
| Gene Symbol: Plxnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108106.1 | NP_001101576.1 | 14 | 2712 | ||
| XM_006242202.2 | XP_006242264.1 | 14 | 2482 | |||
| XM_006242205.2 | XP_006242267.1 | 15 | 4950 | |||
| XM_006242206.2 | XP_006242268.1 | 16 | 2697 | |||
| XM_006242207.2 | XP_006242269.1 | 15 | 2605 | |||
| XM_008765727.1 | XP_008763949.1 | 14 | 2482 | |||
| XM_008765728.1 | XP_008763950.1 | 14 | 4788 | |||
| XM_008765729.1 | XP_008763951.1 | 13 | 2432 | |||

Plxnb2
Genomic Map