siRNA Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079939.1 | NP_001073408.1 | 9 | 1321 | ||
| GenBank mRNA | BC129102.1 | - | 9 | 1321 | 7099869 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmf2| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079939.1 | NP_001073408.1 | 9 | 1321 | ||
| GenBank mRNA | BC129102.1 | - | 9 | 1321 | 7099869 | |