siRNA Details
| Gene Symbol: Arsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034933.2 | NP_001030105.2 | 4, 5 | 855 | ||
| GenBank mRNA | BC105852.1 | - | 4, 5 | 1236 | 7452956 | |
| FQ212178.1 | - | 1817 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arsa| Gene Symbol: Arsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034933.2 | NP_001030105.2 | 4, 5 | 855 | ||
| GenBank mRNA | BC105852.1 | - | 4, 5 | 1236 | 7452956 | |
| FQ212178.1 | - | 1817 | ||||