siRNA Details
| Gene Symbol: Pced1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039454.1 | NP_001034543.1 | 5 | 763 | ||
| XM_008765737.1 | XP_008763959.1 | 2 | 1055 | |||
| XM_008765738.1 | XP_008763960.1 | 2 | 1052 | |||
| XM_008765739.1 | XP_008763961.1 | 2 | 949 | |||
| XM_008765740.1 | XP_008763962.1 | 1 | 414 | |||
| XM_008765741.1 | XP_008763963.1 | 3 | 1325 | |||
| XM_008765742.1 | XP_008763964.1 | 2 | 1702 | |||
| XM_008765743.1 | XP_008763965.1 | 2 | 946 | |||
| XM_008765744.1 | XP_008763966.1 | 2 | 1699 | |||
| GenBank mRNA | BC112322.1 | - | 5 | 763 | 7936449 | |

Pced1b
Genomic Map