siRNA Details
| Gene Symbol: Atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100895.1 | NP_001094365.1 | 1, 2 | 264 | ||
| GenBank mRNA | BC091222.1 | - | 1, 2 | 238 | ||
| BC105853.1 | - | 1, 2 | 261 | |||
| BC168224.1 | - | 1, 2 | 265 | |||
| CO565801.1 | - | 264 | ||||

Atf1
Genomic Map