siRNA Details
| Gene Symbol: Mettl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037355.1 | NP_001032432.1 | 1 | 165 | ||
| GenBank mRNA | BC105760.1 | - | 1 | 165 | 5623503 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl7a| Gene Symbol: Mettl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037355.1 | NP_001032432.1 | 1 | 165 | ||
| GenBank mRNA | BC105760.1 | - | 1 | 165 | 5623503 | |