siRNA Details
| Gene Symbol: Krt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008806.1 | NP_001008806.1 | 3 | 751 | ||
| XM_006242395.2 | XP_006242457.1 | 3 | 818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt4| Gene Symbol: Krt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008806.1 | NP_001008806.1 | 3 | 751 | ||
| XM_006242395.2 | XP_006242457.1 | 3 | 818 | |||