siRNA Details
| Gene Symbol: Hoxc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_235695.7 | XP_235695.3 | 2 | 877 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc13| Gene Symbol: Hoxc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_235695.7 | XP_235695.3 | 2 | 877 | ||