siRNA Details
| Gene Symbol: Smug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177934.3 | NP_808795.1 | 2 | 397 | ||
| XM_006242399.2 | XP_006242461.1 | 1 | 218 | |||
| XM_008765749.1 | XP_008763971.1 | 2 | 274 | |||
| GenBank mRNA | AY191521.1 | - | 2 | 402 | ||

Smug1
Genomic Map