siRNA Details
| Gene Symbol: Copz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108117.1 | NP_001101587.1 | 3, 4 | 213 | ||
| GenBank mRNA | FQ226451.1 | - | 3, 4 | 189 | ||
| FQ234671.1 | - | 3, 4 | 189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copz1| Gene Symbol: Copz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108117.1 | NP_001101587.1 | 3, 4 | 213 | ||
| GenBank mRNA | FQ226451.1 | - | 3, 4 | 189 | ||
| FQ234671.1 | - | 3, 4 | 189 | |||