siRNA Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108120.1 | NP_001101590.1 | 4 | 489 | ||
| XM_006257360.2 | XP_006257422.1 | 5 | 570 | |||
| XM_006257361.2 | XP_006257423.1 | 5 | 513 | |||
| XM_008773417.1 | XP_008771639.1 | 4 | 488 | |||
| GenBank mRNA | BC166561.1 | - | 4 | 425 | 7315205 | |

Nxt2
Genomic Map