siRNA Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145766.2 | NP_665709.2 | 17 | 1942 | ||
| XM_006239527.2 | XP_006239589.1 | 16 | 2498 | |||
| XM_006239528.2 | XP_006239590.1 | 17 | 2305 | |||
| XM_006239529.2 | XP_006239591.1 | 17 | 2226 | |||
| XM_006239530.1 | XP_006239592.1 | 17 | 1926 | |||
| XM_006239531.2 | XP_006239593.1 | 17 | 2030 | |||
| XM_006239532.2 | XP_006239594.1 | 12 | 2374 | |||
| XM_006239533.1 | XP_006239595.1 | 12 | 1351 | |||
| GenBank mRNA | DY544650.1 | - | 5 | 538 | ||

Cdk11b
Genomic Map