siRNA Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009696.1 | NP_001009696.1 | 6 | 716 | ||
| GenBank mRNA | BC087627.1 | - | 6 | 716 | 7318159 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcun1d5| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009696.1 | NP_001009696.1 | 6 | 716 | ||
| GenBank mRNA | BC087627.1 | - | 6 | 716 | 7318159 | |