siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108123.1 | NP_001101593.1 | 11 | 2370 | ||
| XM_006242521.2 | XP_006242583.1 | 12 | 2680 | |||
| XM_006242522.1 | XP_006242584.1 | 11 | 1497 | |||
| XM_008765932.1 | XP_008764154.1 | 12 | 1387 | |||

Mtmr2
Genomic Map