siRNA Details
| Gene Symbol: Sesn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108125.1 | NP_001101595.1 | 4 | 657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sesn3| Gene Symbol: Sesn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108125.1 | NP_001101595.1 | 4 | 657 | ||