siRNA Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270549.1 | NP_001257478.1 | 3 | 875 | ||
| NM_199397.2 | NP_955429.1 | 3 | 875 | |||
| GenBank mRNA | AJ557015.1 | - | 3 | 875 | ||
| GQ499840.1 | - | 3 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Panx1| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270549.1 | NP_001257478.1 | 3 | 875 | ||
| NM_199397.2 | NP_955429.1 | 3 | 875 | |||
| GenBank mRNA | AJ557015.1 | - | 3 | 875 | ||
| GQ499840.1 | - | 3 | 539 | |||