siRNA Details
| Gene Symbol: Tmed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013432.1 | NP_001013450.1 | 4 | 556 | ||
| XM_006242644.1 | XP_006242706.1 | 5 | 2035 | |||
| XM_008765940.1 | XP_008764162.1 | 5 | 1970 | |||
| GenBank mRNA | BC091167.1 | - | 4 | 556 | 7324543 | |

Tmed1
Genomic Map