siRNA Details
Gene Symbol: RGD1309188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108129.1 | NP_001101599.1 | 2 | 920 | ||
GenBank mRNA | FQ213254.1 | - | 2 | 722 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1309188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108129.1 | NP_001101599.1 | 2 | 920 | ||
GenBank mRNA | FQ213254.1 | - | 2 | 722 |