siRNA Details
| Gene Symbol: RGD1309188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108129.1 | NP_001101599.1 | 2 | 962 | ||
| GenBank mRNA | FQ213254.1 | - | 2 | 764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309188| Gene Symbol: RGD1309188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108129.1 | NP_001101599.1 | 2 | 962 | ||
| GenBank mRNA | FQ213254.1 | - | 2 | 764 | ||