siRNA Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108131.1 | NP_001101601.1 | 1 | 105 | ||
| XM_006242747.2 | XP_006242809.1 | 1 | 713 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx19| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108131.1 | NP_001101601.1 | 1 | 105 | ||
| XM_006242747.2 | XP_006242809.1 | 1 | 713 | |||