siRNA Details
| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108132.1 | NP_001101602.1 | 5 | 1209 | ||
| XM_006242720.2 | XP_006242782.1 | 5 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx20| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108132.1 | NP_001101602.1 | 5 | 1209 | ||
| XM_006242720.2 | XP_006242782.1 | 5 | 1204 | |||