siRNA Details
| Gene Symbol: Jam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004269.1 | NP_001004269.1 | 7 | 745 | ||
| XM_008766054.1 | XP_008764276.1 | 6 | 1234 | |||
| GenBank mRNA | BC079429.1 | - | 7 | 745 | 7114667 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jam3| Gene Symbol: Jam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004269.1 | NP_001004269.1 | 7 | 745 | ||
| XM_008766054.1 | XP_008764276.1 | 6 | 1234 | |||
| GenBank mRNA | BC079429.1 | - | 7 | 745 | 7114667 | |