siRNA Details
| Gene Symbol: Arhgap32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242777.2 | XP_006242839.1 | 13 | 1755 | ||
| XM_006242778.2 | XP_006242840.1 | 8 | 4326 | |||
| XM_008766070.1 | XP_008764292.1 | 17 | 2246 | |||
| XM_008766071.1 | XP_008764293.1 | 17 | 2246 | |||
| XM_008766072.1 | XP_008764294.1 | 16 | 2204 | |||
| XM_008766073.1 | XP_008764295.1 | 16 | 2204 | |||
| XM_008766074.1 | XP_008764296.1 | 17 | 3330 | |||
| GenBank mRNA | BC168683.1 | - | 3 | 358 | ||

Arhgap32
Genomic Map