siRNA Details
| Gene Symbol: Fli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017381.1 | NP_001017381.1 | 9 | 1300 | ||
| XM_008766055.1 | XP_008764277.1 | 8 | 1269 | |||
| GenBank mRNA | AY972521.1 | - | 9 | 1300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fli1| Gene Symbol: Fli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017381.1 | NP_001017381.1 | 9 | 1300 | ||
| XM_008766055.1 | XP_008764277.1 | 8 | 1269 | |||
| GenBank mRNA | AY972521.1 | - | 9 | 1300 | ||