siRNA Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108134.2 | NP_001101604.1 | 3 | 1315 | ||
| XM_006242785.1 | XP_006242847.1 | 4 | 1348 | |||
| XM_006242786.2 | XP_006242848.1 | 4 | 1876 | |||
| GenBank mRNA | BC087674.1 | - | 2 | 1720 | ||
| BC158693.1 | - | 3 | 1315 | 9023160 | ||

Pus3
Genomic Map