siRNA Details
Gene Symbol: Robo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108135.1 | NP_001101605.1 | 7 | 1337 | ||
XM_008766057.1 | XP_008764279.1 | 8 | 1462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Robo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108135.1 | NP_001101605.1 | 7 | 1337 | ||
XM_008766057.1 | XP_008764279.1 | 8 | 1462 |