siRNA Details
| Gene Symbol: Panx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199398.1 | NP_955430.1 | 4 | 1159 | ||
| GenBank mRNA | AJ557017.1 | - | 4 | 1159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Panx3| Gene Symbol: Panx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199398.1 | NP_955430.1 | 4 | 1159 | ||
| GenBank mRNA | AJ557017.1 | - | 4 | 1159 | ||