siRNA Details
| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191792.1 | NP_001178721.1 | 12 | 1669 | ||
| XM_006242848.2 | XP_006242910.1 | 12 | 1994 | |||
| XM_006242850.2 | XP_006242912.1 | 12 | 2243 | |||
| XM_008766123.1 | XP_008764345.1 | 12 | 1997 | |||
| GenBank mRNA | FQ221553.1 | - | 12 | 1920 | ||
| FQ223998.1 | - | 12 | 1920 | |||

Ubash3b
Genomic Map