siRNA Details
| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014089.1 | NP_001014111.1 | 5 | 905 | ||
| XM_006242859.2 | XP_006242921.1 | 5 | 3552 | |||
| XM_008766134.1 | XP_008764356.1 | 6 | 3515 | |||
| GenBank mRNA | BC087163.1 | - | 5 | 905 | 7135124 | |
| FQ230557.1 | - | 3 | 249 | |||

Tbcel
Genomic Map