siRNA Details
| Gene Symbol: Tmem136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108136.1 | NP_001101606.1 | 3 | 598 | ||
| GenBank mRNA | FQ212295.1 | - | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem136| Gene Symbol: Tmem136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108136.1 | NP_001101606.1 | 3 | 598 | ||
| GenBank mRNA | FQ212295.1 | - | 600 | |||