siRNA Details
| Gene Symbol: Mfrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108137.2 | NP_001101607.1 | 6 | 878 | ||
| XM_006242933.2 | XP_006242995.1 | 6 | 1323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfrp| Gene Symbol: Mfrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108137.2 | NP_001101607.1 | 6 | 878 | ||
| XM_006242933.2 | XP_006242995.1 | 6 | 1323 | |||