siRNA Details
| Gene Symbol: Nlrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025010.1 | NP_001020181.1 | 6 | 977 | ||
| GenBank mRNA | BC089834.1 | - | 6 | 977 | 7325650 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrx1| Gene Symbol: Nlrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025010.1 | NP_001020181.1 | 6 | 977 | ||
| GenBank mRNA | BC089834.1 | - | 6 | 977 | 7325650 | |