siRNA Details
| Gene Symbol: Cd3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108140.1 | NP_001101610.1 | 5 | 435 | ||
| XM_008766161.1 | XP_008764383.1 | 5 | 1037 | |||
| GenBank mRNA | FQ228137.1 | - | 5 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd3e| Gene Symbol: Cd3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108140.1 | NP_001101610.1 | 5 | 435 | ||
| XM_008766161.1 | XP_008764383.1 | 5 | 1037 | |||
| GenBank mRNA | FQ228137.1 | - | 5 | 457 | ||