siRNA Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108142.1 | NP_001101612.1 | 3 | 1139 | ||
| XM_006242941.2 | XP_006243003.1 | 3 | 1670 | |||
| XM_006242942.2 | XP_006243004.1 | 3 | 1670 | |||
| XM_006242943.2 | XP_006243005.1 | 3 | 1669 | |||

Sidt2
Genomic Map