siRNA Details
| Gene Symbol: Usp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108144.1 | NP_001101614.1 | 12 | 1211 | ||
| XM_006243006.2 | XP_006243068.1 | 12 | 1320 | |||
| XM_006243007.2 | XP_006243069.1 | 12 | 1317 | |||
| XM_006243008.2 | XP_006243070.1 | 12 | 1320 | |||
| XM_006243009.2 | XP_006243071.1 | 12 | 1320 | |||
| XM_006243010.2 | XP_006243072.1 | 12 | 1317 | |||
| XM_006243011.2 | XP_006243073.1 | 12 | 1320 | |||
| XM_006243012.2 | XP_006243074.1 | 12 | 1317 | |||
| GenBank mRNA | AB240643.1 | - | 12 | 1220 | ||

Usp28
Genomic Map