siRNA Details
| Gene Symbol: Bco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127712.1 | NP_001121184.1 | 8 | 1000 | ||
| GenBank mRNA | AY325176.1 | - | 772 | |||
| BC133725.1 | - | 8 | 1000 | 7100438 | ||
| DQ083174.1 | - | 6 | 664 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bco2| Gene Symbol: Bco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127712.1 | NP_001121184.1 | 8 | 1000 | ||
| GenBank mRNA | AY325176.1 | - | 772 | |||
| BC133725.1 | - | 8 | 1000 | 7100438 | ||
| DQ083174.1 | - | 6 | 664 | |||