siRNA Details
| Gene Symbol: Pih1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191793.1 | NP_001178722.1 | 7 | 1222 | ||
| XM_006243028.2 | XP_006243090.1 | 6 | 988 | |||
| XM_006243029.2 | XP_006243091.1 | 5 | 895 | |||
| XM_006243030.2 | XP_006243092.1 | 5 | 894 | |||

Pih1d2
Genomic Map