siRNA Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005889.2 | NP_001005889.2 | 8 | 942 | ||
| GenBank mRNA | AY428868.1 | - | 7 | 767 | ||
| BC087147.1 | - | 8 | 942 | 7130553 | ||
| FQ218538.1 | - | 5 | 611 | |||
| KF307742.1 | - | 7 | 785 | |||

Rdx
Genomic Map