siRNA Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005889.2 | NP_001005889.2 | 8, 9 | 1168 | ||
| GenBank mRNA | AY428868.1 | - | 7, 8 | 993 | ||
| BC087147.1 | - | 8, 9 | 1168 | 7130553 | ||
| FQ218538.1 | - | 5, 6 | 837 | |||
| KF307742.1 | - | 7, 8 | 1011 | |||

Rdx
Genomic Map